Citric acid monohydrate, 99.5+%
Citric acid monohydrate, 99.5+%
Citric acid monohydrate, 99.5+%
Citric acid monohydrate, 99.5+%
Thermo Scientific Chemicals
Citric acid monohydrate, 99.5+%
CAS: 5949-29-1 | C6H10O8 | 210.138 g/mol
Have Questions?
Catalog NumberQuantity
ALF022869.36500 g
Catalog number ALF022869.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCitric acid monohydrate
CAS5949-29-1
Health Hazard 1H319-H500
ColorWhite
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, flavoring agent and chelating agent.