Crystal Violet, ACS, 90+%
Crystal Violet, ACS, 90+%
Crystal Violet, ACS, 90+%
Crystal Violet, ACS, 90+%
Thermo Scientific Chemicals
Crystal Violet, ACS, 90+%
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Have Questions?
Catalog NumberQuantity
ALF022866.1425 g
Catalog number ALF022866.14
Price (MYR)
338.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
338.00
EA
Add to cart
Specifications
Chemical Name or MaterialCrystal Violet
Melting Point∼215°C (decomposition)
CAS548-62-9
Health Hazard 1H302-H318-H351
Health Hazard 2GHS H Statement
H318-H351-H302
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
View more
Crystal Violet is used as an acid-base indicator, an alcohol denaturant, a biological stain, a textile dye and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.