Cal-Paks™ Weight Sets: Class 1
Cal-Paks™ Weight Sets: Class 1
Actual product may vary
Thermo Scientific Chemicals
Cal-Paks™ Weight Sets: Class 1
Highly polished Alloy 8 stainless-steel weights
Have Questions?
Catalog NumberMassWeight (Metric)Includes
ALF022156.1820 mg, 2 g, 20 g22.0 gCylindrical weight: 20g, 2g, sheet weight: 20mg, Polycarbonate case; NVLAP certificate
Catalog number ALF022156.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Mass:
20 mg, 2 g, 20 g
Weight (Metric):
22.0 g
Includes:
Cylindrical weight: 20g, 2g, sheet weight: 20mg, Polycarbonate case; NVLAP certificate
Highly polished Alloy 8 stainless-steel weights
ANSI™/ASTM™ Class 1.
  • Components and case are linked through the use of a unique serial number
  • Clear plastic case
  • Individual weights in high-quality polycarbonate cases
Specifications
Product TypeSmall Weight Set
Certifications/ComplianceNVLAP, ISO9001
ClassificationASTM Class 1
Weight (Metric)22.0 g
Density7.95g/cm3 at 20°C
Mass20 mg, 2 g, 20 g
IncludesCylindrical weight: 20g, 2g, sheet weight: 20mg, Polycarbonate case; NVLAP certificate
Quantity3/Set
Packaging Quantity3
Unit SizeEA