Cerium(III) 2,4-pentanedionate hydrate
Cerium(III) 2,4-pentanedionate hydrate
Cerium(III) 2,4-pentanedionate hydrate
Cerium(III) 2,4-pentanedionate hydrate
Thermo Scientific Chemicals
Cerium(III) 2,4-pentanedionate hydrate
CAS: 206996-61-4 | C15H21CeO6 | 437.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF021116.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(III) 2,4-pentanedionate hydrate
Melting Point131°C to 132°C
CAS206996-61-4
ColorRed to Yellow
Health Hazard 1H315-H319-H335
View more
Cerium(III) 2,4-pentanedionate hydrate used as pharmaceutical intermediate.