Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulfonate
Thermo Scientific Chemicals
Sodium trifluoromethanesulfonate
CAS: 2926-30-9 | CF3NaO3S | 172.053 g/mol
Have Questions?
Catalog NumberQuantity
ALF020837.042 g
Catalog number ALF020837.04
Price (MYR)
254.00
EA
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (MYR)
254.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium trifluoromethanesulfonate
CAS2926-30-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.