Tetraethylene glycol dimethyl ether, 98+%
Tetraethylene glycol dimethyl ether, 98+%
Tetraethylene glycol dimethyl ether, 98+%
Tetraethylene glycol dimethyl ether, 98+%
Tetraethylene glycol dimethyl ether, 98+%
Thermo Scientific Chemicals
Tetraethylene glycol dimethyl ether, 98+%
CAS: 143-24-8 | C10H22O5 | 222.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF020443.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylene glycol dimethyl ether
CAS143-24-8
Health Hazard 1H360Df
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
Tetraethylene glycol dimethyl ether is used in lithium-ion battery technology. It is associated with trifluoroethanol and used as a working pair for organic absorption heat pumps. It acts as a solvent for cleaning and degreasing due to its chemical and thermal stability. Moreover, it is suitable solvent for high temperature reactions due its high boiling point. Further, it is used in the selective adsorption of proteins during the promotion of cell adhesion.