2-Propanol, Semiconductor Grade, 99.5% min
2-Propanol, Semiconductor Grade, 99.5% min
2-Propanol, Semiconductor Grade, 99.5% min
2-Propanol, Semiconductor Grade, 99.5% min
Thermo Scientific Chemicals
2-Propanol, Semiconductor Grade, 99.5% min
Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog NumberQuantity
ALF019397.K21 L
Catalog number ALF019397.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material2-Propanol
CAS67-63-0
Health Hazard 1H225-H319-H335-H336-H373
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
Melting Point°C to 90°C
View more
2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.