Methanol, Semiconductor Grade, 99.9% min
Methanol, Semiconductor Grade, 99.9% min
Methanol, Semiconductor Grade, 99.9% min
Methanol, Semiconductor Grade, 99.9% min
Thermo Scientific Chemicals
Methanol, Semiconductor Grade, 99.9% min
Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantity
ALF019393.K21 L
Catalog number ALF019393.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Melting Point°C to 98°C
CAS67-56-1
ColorColorless
Health Hazard 1H225-H301+H311+H331-H370-H372
View more
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.