1,2-Dichlorobenzene, HPLC Grade, 98% min
1,2-Dichlorobenzene, HPLC Grade, 98% min
1,2-Dichlorobenzene, HPLC Grade, 98% min
1,2-Dichlorobenzene, HPLC Grade, 98% min
Thermo Scientific Chemicals
1,2-Dichlorobenzene, HPLC Grade, 98% min
CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol
Have Questions?
Catalog NumberQuantity
ALF019385.K74 L
Catalog number ALF019385.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or Material1,2-Dichlorobenzene
CAS95-50-1
Health Hazard 1H227-H302+H332-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
View more
1,2-Dichlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.