Selenic acid, 40% aq. soln.
Selenic acid, 40% aq. soln.
Selenic acid, 40% aq. soln.
Selenic acid, 40% aq. soln.
Thermo Scientific Chemicals
Selenic acid, 40% aq. soln.
CAS: 7783-08-6 | K2O4Se | 221.16 g/mol
Have Questions?
Catalog NumberQuantity
ALF018851.22100 g
Catalog number ALF018851.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSelenic acid
CAS7783-08-6
ColorColorless
Health Hazard 1H301+H331-H373
Health Hazard 2GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
View more
Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.