Allyl methyl ether, 94%
Allyl methyl ether, 94%
Allyl methyl ether, 94%
Allyl methyl ether, 94%
Thermo Scientific Chemicals
Allyl methyl ether, 94%
CAS: 627-40-7 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF018652.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAllyl methyl ether
CAS627-40-7
Health Hazard 1H225
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
Allyl methyl ether it used as pharmaceutical intermediate. 