Gallium Tin alloy, 99.99% (metals basis)
Gallium Tin alloy, 99.99% (metals basis)
Gallium Tin alloy, 99.99% (metals basis)
Gallium Tin alloy, 99.99% (metals basis)
Thermo Scientific Chemicals
Gallium Tin alloy, 99.99% (metals basis)
CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Catalog NumberQuantity
ALF018161.0910 g
Catalog number ALF018161.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGallium Tin alloy
ColorSilver
CAS7440-31-5
Melting Point≥25°C
Health Hazard 1H290-H314-H335
View more
Gallium Tin alloy is used to cool computer chips in place of water.  Used in medical thermometers.