Methyl Orange, ACS
Methyl Orange, ACS
Methyl Orange, ACS
Thermo Scientific Chemicals
Methyl Orange, ACS
CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol
Have Questions?
Catalog NumberQuantity
ALF017874.065 g
Catalog number ALF017874.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl Orange
Melting Point>300°C
CAS547-58-0
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
View more
Methyl orange is a pH indicator frequently used in titrations, also used for histological microscopy.