Bis(2-ethylhexyl) phosphate, 95%
Bis(2-ethylhexyl) phosphate, 95%
Bis(2-ethylhexyl) phosphate, 95%
Bis(2-ethylhexyl) phosphate, 95%
Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%
Extraction of metals | CAS: 298-07-7 | C16H35O4P | 322.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number ALF017723.A3
Price (MYR)
2,522.00
EA
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
2,522.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(2-ethylhexyl) phosphate
CAS298-07-7
Health Hazard 1H302-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
Extraction of metals.Bis(2-ethylhexyl) phosphate is used as a lubricant additive, corrosion inhibitor and a metal extractant. It is involved in the solvent extraction of uranium salts and rare earth metals. Iron(II) ions are extracted after reduction of Iron(III) ions. Further, it is used as a plasticizer and as a solvent in the synthesis of plastic.