Bismuth(III) chloride, anhydrous, 99.9% (metals basis)
Bismuth(III) chloride, anhydrous, 99.9% (metals basis)
Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.9% (metals basis)
CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Catalog NumberQuantity
ALF017115.1425 g
Catalog number ALF017115.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Pointca 430°C (sublimation)
Chemical Name or MaterialBismuth(III) chloride
Melting Point230°C to 232°C
CAS7787-60-2
ColorWhite
View more
Catalysts for organic reactions, preparation of Bi salts.Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.