Amberlite™ IRN-77, ion exchange resin, nuclear grade
Amberlite™ IRN-77, ion exchange resin, nuclear grade
Thermo Scientific Chemicals
Amberlite™ IRN-77, ion exchange resin, nuclear grade
Styrene-DVB; Strongly acidic cation exchange resin | CAS: 11128-94-2 | Styrene-DVB
Have Questions?
Catalog NumberQuantity
ALF016821.36500 g
Catalog number ALF016821.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite IRN-77
Physical FormBeads
CAS11128-94-2
Recommended StorageAmbient temperatures
TSCAYes
View more
Amberlite™ IRN-77 is used in the primary water treatment for the removal of fission products, activated corrosion products and suspended matter. It is also used to control the pH of the reactor coolant stream by removing the excess lithium. Further, it is used in the removal of radioactive cations such as Cesium from waste streams. In addition to this, it is used as a cation exchanger to determine the sorption activity towards cesium(I) and strontium(II) using isothermal titration calorimetry and solution-depletion method.