Trimethylolpropane triacrylate, stabilized
Trimethylolpropane triacrylate, stabilized
Trimethylolpropane triacrylate, stabilized
Trimethylolpropane triacrylate, stabilized
Thermo Scientific Chemicals
Trimethylolpropane triacrylate, stabilized
CAS: 15625-89-5 | C15H20O6 | 296.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF016445.36
Price (MYR)
618.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
618.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrimethylolpropane triacrylate
Name NoteStabilized
CAS15625-89-5
Health Hazard 1H227-H315-H317-H319
Health Hazard 2GHS H Statement
H315-H319-H317-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.