Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
Thermo Scientific Chemicals
Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
CAS: 1330-20-7 | (CH3)2C6H4
Have Questions?
Catalog NumberQuantity
ALF016371.K21 L
Catalog number ALF016371.K2
Price (MYR)
351.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
351.00
EA
Add to cart
Specifications
Chemical Name or MaterialXylenes
Melting Point<°C to 25°C
Name NoteFor ACS analysis
CAS1330-20-7
ColorColorless
View more
Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate.