Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Thermo Scientific Chemicals
Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
CAS: 79-41-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
250 g
Catalog number ALF015546.A5
Price (MYR)
-
Quantity:
3 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialMethacrylic acid
Name NoteStabilized with 250ppm 4-methoxyphenol
CAS79-41-4
Health Hazard 1H227-H302+H332-H311-H314-H335
Health Hazard 2GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
View more
Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly (methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.