Tantalum(V) oxide, 99% (metals basis)
Tantalum(V) oxide, 99% (metals basis)
Tantalum(V) oxide, 99% (metals basis)
Tantalum(V) oxide, 99% (metals basis)
Thermo Scientific Chemicals
Tantalum(V) oxide, 99% (metals basis)
CAS: 1314-61-0 | O5Ta2 | 441.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALF014709.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTantalum(V) oxide
Melting Point1800°C
CAS1314-61-0
Health Hazard 1H500
Recommended StorageAmbient temperatures
View more
Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.