Nickel(II) chloride, anhydrous, 99% (metals basis)
Nickel(II) chloride, anhydrous, 99% (metals basis)
Nickel(II) chloride, anhydrous, 99% (metals basis)
Nickel(II) chloride, anhydrous, 99% (metals basis)
Thermo Scientific Chemicals
Nickel(II) chloride, anhydrous, 99% (metals basis)
CAS: 7718-54-9 | Cl2Ni | 129.593 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF014687.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel(II) chloride, anhydrous
Melting Point1001°C
CAS7718-54-9
ColorYellow to Orange
Health Hazard 1H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500
View more
Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.