Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals
Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals
Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals
Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF014660.36500 g
Catalog number ALF014660.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point238°C
Chemical Name or MaterialAmmonium hydrogen fluoride
Melting Point120°C to 122°C
CAS1341-49-7
Health Hazard 1H301-H314-H335
View more
Ammonium hydrogen fluoride is used as a chemical reagent in analysis and an antiseptic. It is also used in ceramics, electroplating, component of glass etchants as well as food processing equipment disinfectant. It acts as an intermediate in the production of hydrofluoric acid from hexafluorosilicic acid.