Antimony powder, -200 mesh, 99.999% (metals basis)
Antimony powder, -200 mesh, 99.999% (metals basis)
Antimony powder, -200 mesh, 99.999% (metals basis)
Antimony powder, -200 mesh, 99.999% (metals basis)
Thermo Scientific Chemicals
Antimony powder, -200 mesh, 99.999% (metals basis)
CAS: 7440-36-0 | Sb | 121.76 g/mol
Have Questions?
Catalog NumberQuantity
ALF014640.1425 g
Catalog number ALF014640.14
Price (MYR)
2,285.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,285.00
EA
Add to cart
Specifications
Chemical Name or MaterialAntimony
ColorGray
CAS7440-36-0
Health Hazard 1H335-H351-H373
AppearanceGray powder