Samarium(III) acetate hydrate, 99.9% (REO)
Samarium(III) acetate hydrate, 99.9% (REO)
Samarium(III) acetate hydrate, 99.9% (REO)
Samarium(III) acetate hydrate, 99.9% (REO)
Samarium(III) acetate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Samarium(III) acetate hydrate, 99.9% (REO)
CAS: 17829-86-6 | C6H9O6Sm | 327.49 g/mol
Have Questions?
Catalog NumberQuantity
ALF014604.22100 g
Catalog number ALF014604.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point300°C
Chemical Name or MaterialSamarium(III) acetate hydrate
CAS17829-86-6
Recommended StorageAmbient temperatures
Physical FormCrystalline Aggregates
View more
Samarium Acetate has specialized uses in glass, phosphors, lasers, and thermoelectric devices. Another important application of Samarium and its compounds is as catalyst and chemical reagent. One of the most important applications of Samarium is in Samarium-Cobalt magnets. . These magnets are found in small motors, headphones, and high-end magnetic pickups for guitars and related musical instruments.