Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)
Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)
Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)
Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)
CAS: 14483-18-2 | H10HoN3O14 | 441.017 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF014588.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHolmium(III) nitrate pentahydrate
CAS14483-18-2
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Holmium(III) nitrate pentahydrate is used to prepare holmium-beta-cyclodextrin complex by reacting with beta- cyclodextrin, which posses potential pharmacological applications.