Ammonium formate, 98+%
Ammonium formate, 98+%
Ammonium formate, 98+%
Ammonium formate, 98+%
Thermo Scientific Chemicals
Ammonium formate, 98+%
CAS: 540-69-2 | CH5NO2 | 63.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF014517.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium formate
Melting Point∼116°C
CAS540-69-2
ColorWhite
Health Hazard 1H315-H319-H335-H500
View more
Ammonium formate is widely used in various organic reactions like Leuckart reaction which involves the reductive amination of aldehydes and ketones. It serves as a buffer in high performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC/MS). It finds application in palladium on carbon (Pd/C) reduction of functional groups. For example, reduction of alkenes to alkanes and formaldehyde to methanol. It is also used to prepare formic acid insitu as well as used to store formic acid by making it as an ammonium salt.