Ammonium hydrogen sulfate, 99.9% (metals basis)
Ammonium hydrogen sulfate, 99.9% (metals basis)
Ammonium hydrogen sulfate, 99.9% (metals basis)
Ammonium hydrogen sulfate, 99.9% (metals basis)
Thermo Scientific Chemicals
Ammonium hydrogen sulfate, 99.9% (metals basis)
CAS: 7803-63-6 | H5NO4S | 115.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF014485.22100 g
Catalog number ALF014485.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hydrogen sulfate
Melting Point146.9°C
CAS7803-63-6
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.