Ammonium hydrogen carbonate, 99.0%
Ammonium hydrogen carbonate, 99.0%
Ammonium hydrogen carbonate, 99.0%
Ammonium hydrogen carbonate, 99.0%
Ammonium hydrogen carbonate, 99.0%
Thermo Scientific Chemicals
Ammonium hydrogen carbonate, 99.0%
CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol
Have Questions?
Catalog NumberQuantity
ALF014249.A11 kg
Catalog number ALF014249.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hydrogen carbonate
Melting Point∼60°C (decomposition)
CAS1066-33-7
ColorWhite
Health Hazard 1H302
View more
In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition.Ammonium hydrogen carbonate is used as a reagent in chemical industries. It is an active component used in the manufacturing of fire-extinguishing compounds, pharmaceuticals, dyes, pigments and fertilizers. It finds application in the plastic, ceramics, leather and rubber industries. It is also used in cooling baths, scale and gypsum removing agents, leavening agent for baked goods and a defatting agent for textiles. Further, it acts as a buffer used in the purification of chemicals. In addition to this, it is involved in the gel digestion of proteins by trypsin and mass spectrometric analysis of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments
  • Also used for compost treatment to accelerate decomposition
RUO – Research Use Only