Iodine, crystalline, 99.5%
Iodine, crystalline, 99.5%
Iodine, crystalline, 99.5%
Iodine, crystalline, 99.5%
Thermo Scientific Chemicals
Iodine, crystalline, 99.5%
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Catalog NumberQuantity
ALF014248.30250 g
Catalog number ALF014248.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIodine
Melting Point183°C to 186° (sublimation)
CAS7553-56-2
Health Hazard 1H302+H312+H332-H315-H319-H335-H372
Health Hazard 3P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c