Zinc, Oil based standard solution, Specpure™ Zn 1000μg/g
Zinc, Oil based standard solution, Specpure™ Zn 1000μg/g
Thermo Scientific Chemicals
Zinc, Oil based standard solution, Specpure™ Zn 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014200.1850 g
Catalog number ALF014200.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc Oil based standard solution
Name NoteZn 1000μg/g
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c