Telluric acid dihydrate, 99+%
Telluric acid dihydrate, 99+%
Telluric acid dihydrate, 99+%
Telluric acid dihydrate, 99+%
Thermo Scientific Chemicals
Telluric acid dihydrate, 99+%
CAS: 13451-14-4 | H6O6Te | 229.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF014197.18
Price (MYR)
1,397.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,397.00
EA
Add to cart
Specifications
Boiling Point160°C (decomposition)
Chemical Name or MaterialTelluric acid dihydrate
Melting Point136°C
CAS13451-14-4
ColorWhite
View more
Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.