Platinum wire, 0.762mm (0.03in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.762mm (0.03in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.762mm (0.03in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.762mm (0.03in) dia, hard, 99.95% (metals basis)
Thermo Scientific Chemicals
Platinum wire, 0.762mm (0.03in) dia, hard, 99.95% (metals basis)
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF014195.BW50 cm
Catalog number ALF014195.BW
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum wire
CAS7440-06-4
Recommended StorageAmbient temperatures
EINECS Number231-116-1
TSCAYes