Tri-n-butylphosphine, 94%
Tri-n-butylphosphine, 94%
Tri-n-butylphosphine, 94%
Tri-n-butylphosphine, 94%
Thermo Scientific Chemicals
Tri-n-butylphosphine, 94%
CAS: 998-40-3 | C12H27P | 202.32 g/mol
Have Questions?
Catalog NumberQuantity
ALF014114.1850 g
Catalog number ALF014114.18
Price (MYR)
376.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
376.00
EA
Add to cart
Specifications
Chemical Name or MaterialTri-n-butylphosphine
CAS998-40-3
Health Hazard 1H226-H250-H302+H312-H314-H335
Health Hazard 2GHS H Statement
H250-H252-H314-H318-H302-H312
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P210-P222-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Tri-n-butylphosphine is used as a catalyst modifier in the cobalt-catalyzed hydroformylation of alkenes for industrial use. It is also used as a strong reducing agent and polymerization cross-linking catalyst. It is a used as an intermediate for phase transfer catalysts. Used with platinum (II) or (IV) in an intermolecular hydroamination of inactivated alkenes. It is used as 1,4-addition catalyst; used with disulfides for the thioetherification of alcohols; acylation catalyst; used to prepare active esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels
RUO – Research Use Only