Tetraethoxysilane, 99+%
Tetraethoxysilane, 99+%
Tetraethoxysilane, 99+%
Tetraethoxysilane, 99+%
Thermo Scientific Chemicals
Tetraethoxysilane, 99+%
As a binding agent and as a polymerization catalyst | CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol
Have Questions?
Catalog NumberQuantity
ALF014082.A42.5 kg
Catalog number ALF014082.A4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Request bulk or custom format
Specifications
Boiling Point168°C
Chemical Name or MaterialTetraethoxysilane
Melting Point-85°C
CAS78-10-4
ColorColorless
View more
As a binding agent and as a polymerization catalystTetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as a cross linking agent in silicon polymers and in aerogel preparations.