Tetraethoxysilane, 99+%
Tetraethoxysilane, 99+%
Tetraethoxysilane, 99+%
Tetraethoxysilane, 99+%
Thermo Scientific Chemicals
Tetraethoxysilane, 99+%
CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol
Have Questions?
Catalog NumberQuantity
ALF014082.36500 g
Catalog number ALF014082.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethoxysilane
CAS78-10-4
Health Hazard 1H226-H319-H332-H335
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
Melting Point85°C
View more
As a binding agent and as a polymerization catalystTetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as a cross linking agent in silicon polymers and in aerogel preparations.