Vanadium(III) chloride, 99% (metals basis)
Vanadium(III) chloride, 99% (metals basis)
Vanadium(III) chloride, 99% (metals basis)
Vanadium(III) chloride, 99% (metals basis)
Thermo Scientific Chemicals
Vanadium(III) chloride, 99% (metals basis)
CAS: 7718-98-1 | Cl3V | 157.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF014039.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialVanadium(III) chloride
CAS7718-98-1
Health Hazard 1H302-H314-H335
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
Recommended StorageAmbient temperatures; Store under Argon
View more
Vanadium(III) chloride acts as an intermediate in organic synthesis and in the manufacture of organic light emitting diodes. It acts as a precursor to other vanadium(III) complexes. Further, it is used in the preparation of vanadium dichloride and organovanadium compounds. In addition to this, it serves as a catalyst in the production of ethylene propylene diene monomer rubber and polyethylene.