Iron(II) sulfide, 99.9% (metals basis)
Iron(II) sulfide, 99.9% (metals basis)
Iron(II) sulfide, 99.9% (metals basis)
Iron(II) sulfide, 99.9% (metals basis)
Thermo Scientific Chemicals
Iron(II) sulfide, 99.9% (metals basis)
CAS: 1317-37-9 | FeS | 87.91 g/mol
Have Questions?
Catalog NumberQuantity
ALF014024.0910 g
Catalog number ALF014024.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIron(II) sulfide
Melting Point1195°C
CAS1317-37-9
ColorBrown to Black
Health Hazard 1Contact with acids liberates toxic gas.
View more
Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.