Potassium tin(IV) oxide trihydrate, 95%
Potassium tin(IV) oxide trihydrate, 95%
Potassium tin(IV) oxide trihydrate, 95%
Potassium tin(IV) oxide trihydrate, 95%
Thermo Scientific Chemicals
Potassium tin(IV) oxide trihydrate, 95%
CAS: 12125-03-0 | H6K2O6Sn | 298.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF014015.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium tin(IV) oxide trihydrate
Melting Point140°C (decomposition)
CAS12125-03-0
Recommended StorageAmbient temperatures
Physical FormCrystalline
View more
Potassium tin(IV) oxide is a biochemical for proteomics research.