Germanium(II) iodide, 99.99% (metals basis)
Germanium(II) iodide, 99.99% (metals basis)
Germanium(II) iodide, 99.99% (metals basis)
Germanium(II) iodide, 99.99% (metals basis)
Thermo Scientific Chemicals
Germanium(II) iodide, 99.99% (metals basis)
CAS: 13573-08-5 | GeI2 | 326.439 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF014013.06
Price (MYR)
5,892.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
5,892.00
EA
Add to cart
Specifications
Chemical Name or MaterialGermanium(II) iodide
CAS13573-08-5
ColorYellow to Brown
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
Germanium(II) iodide acts as a reagent that forms stable ionic adducts with carbenes. It is also used in the preparation of allylgermanes by reacting with allylstannanes. Further, it reacts with allyl bromide in the presence of zinc iodide to get allylgermanium(IV), which on reacts with carbonyl compounds to give the corresponding homoallylic alcohols.