Germanium(IV) iodide, 99.999% (metals basis)
Germanium(IV) iodide, 99.999% (metals basis)
Germanium(IV) iodide, 99.999% (metals basis)
Germanium(IV) iodide, 99.999% (metals basis)
Thermo Scientific Chemicals
Germanium(IV) iodide, 99.999% (metals basis)
CAS: 13450-95-8 | GeI4 | 580.248 g/mol
Have Questions?
Catalog NumberQuantity
ALF014009.0910 g
Catalog number ALF014009.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGermanium(IV) iodide
Melting Point146°C
CAS13450-95-8
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
Germanium(IV) iodide is a biochemical for proteomics research. It is also used as a primary and secondary intermediate.