Aluminum isopropoxide, 98+%
Aluminum isopropoxide, 98+%
Aluminum isopropoxide, 98+%
Aluminum isopropoxide, 98+%
Thermo Scientific Chemicals
Aluminum isopropoxide, 98+%
Reagent for the reduction of carbonyl compounds | CAS: 555-31-7 | C9H21AlO3 | 204.246 g/mol
Have Questions?
Catalog NumberQuantity
ALF014007.A11 kg
Catalog number ALF014007.A1
Price (MYR)
486.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
486.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum isopropoxide
CAS555-31-7
Health Hazard 1H228-H319-H336
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
View more
Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates. Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Reagent for the reduction of carbonyl compounds
  • Used in alcoholysis and ester exchange reactions
  • Also used in synthesis of higher alkoxides, chelates, and acrylates
RUO – Research Use Only