Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Thermo Scientific Chemicals
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF013990.36500 g
Catalog number ALF013990.36
Price (MYR)
446.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
446.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper powder
CAS7440-50-8
Health Hazard 1H228-H315-H319-H335-H500
Health Hazard 2GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c