Barium, plasma standard solution, Specpure™ Ba 1000μg/mL
Barium, plasma standard solution, Specpure™ Ba 1000μg/mL
Barium, plasma standard solution, Specpure™ Ba 1000μg/mL
Barium, plasma standard solution, Specpure™ Ba 1000μg/mL
Thermo Scientific Chemicals
Barium, plasma standard solution, Specpure™ Ba 1000μg/mL
CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
Catalog NumberQuantity
ALF013876.AE100 mL
Catalog number ALF013876.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBarium
CAS10022-31-8
Health Hazard 1H290-H314-H335
CAS7732-18-5
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c