Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL
Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL
Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL
Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL
Thermo Scientific Chemicals
Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL
CAS: 1314-36-9 | O3Y2 | 225.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number ALF013855.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialYttrium
CAS1314-36-9
Health Hazard 1H290-H314-H335
Health Hazard 2Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
CAS7732-18-5
View more
Yttrium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.