Lead, plasma standard solution, Specpure™ Pb 1000μg/mL
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL
Thermo Scientific Chemicals
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL
CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Catalog NumberQuantity
ALF013853.AE100 mL
Catalog number ALF013853.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialLead
CAS7439-92-1
Health Hazard 1H290-H314-H335-H350-H360-H373
CAS7697-37-2
Health Hazard 3P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Lead, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.