Silver, plasma standard solution, Specpure™ Ag 1000μg/mL
Silver, plasma standard solution, Specpure™ Ag 1000μg/mL
Silver, plasma standard solution, Specpure™ Ag 1000μg/mL
Silver, plasma standard solution, Specpure™ Ag 1000μg/mL
Thermo Scientific Chemicals
Silver, plasma standard solution, Specpure™ Ag 1000μg/mL
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF013849.AE100 mL
Catalog number ALF013849.AE
Price (MYR)
214.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
214.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
Name Noteplasma standard solution
CAS7440-22-4
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c