Sulfur, plasma standard solution, Specpure™ S 1000μg/mL
(NH4)2SO4 in H2O
Have Questions?
Change view
Quantity:
500 mL
100 mL
Catalog number ALF013842.AP
Price (MYR)
740.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
740.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfur, plasma standard solution
Recommended StorageAmbient temperatures
TSCAYes
Vapor Pressure23 hPa (17mm Hg) at 20°C
Physical FormLiquid