Lithium iodide, ultra dry, 99.999% (metals basis)
Lithium iodide, ultra dry, 99.999% (metals basis)
Lithium iodide, ultra dry, 99.999% (metals basis)
Lithium iodide, ultra dry, 99.999% (metals basis)
Lithium iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals
Lithium iodide, ultra dry, 99.999% (metals basis)
CAS: 10377-51-2 | ILi | 133.84 g/mol
Have Questions?
Catalog NumberQuantity
ALF013600.1425 g
Catalog number ALF013600.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point1170°C to 1190°C
Chemical Name or MaterialLithium iodide
Melting Point449°C
Name NoteUltra dry
CAS10377-51-2
View more
It is used as an absorbent in refrigeration process, as a catalyst, and in photography. It is also useful for organic synthesis, pessat iodination, cleavage of esters, ethers, epoxides, decarboethoxylation, carbonylation, and aldol condensation. It is used for manufacturing medicines and mineral waters. It is used in electochemicstry and redox reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only