Sodium chromate tetrahydrate, Reagent Grade
Sodium chromate tetrahydrate, Reagent Grade
Sodium chromate tetrahydrate, Reagent Grade
Sodium chromate tetrahydrate, Reagent Grade
Thermo Scientific Chemicals
Sodium chromate tetrahydrate, Reagent Grade
For Use As Corrosion Inhibitor, in Pigment Manufacture | CAS: 10034-82-9 | CrH8Na2O8 | 234.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF013453.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium chromate tetrahydrate
CAS10034-82-9
ColorYellow
Health Hazard 1H301-H312-H314-H317-H330-H334-H335-H340-H350-H360-H372
Health Hazard 2Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
View more
Corrosion inhibitor, pigment manufactureSodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.