Potassium oxalate monohydrate, ACS, 98.5-101.0%
Potassium oxalate monohydrate, ACS, 98.5-101.0%
Potassium oxalate monohydrate, ACS, 98.5-101.0%
Potassium oxalate monohydrate, ACS, 98.5-101.0%
Thermo Scientific Chemicals
Potassium oxalate monohydrate, ACS, 98.5-101.0%
CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF013452.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium oxalate monohydrate
CAS6487-48-5
Health Hazard 1H302+H312
Health Hazard 2GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
View more
Potassium oxalate monohydrate is used as a chelating agent; a purifying agent in pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, a rust remover for metal treatment, a bleaching agent in textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as a reducing agent in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.